accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 635305-48-5

IUPAC Name: 4,4,5,5-tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(4-methylthiophen-2-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 224.13
Molecular Formula: C11H17BO2S
InChi Key: FNRMPXJVCSMVMA-UHFFFAOYSA-N
SMILES: CC1=CSC(=C1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results