CAS RN 635712-44-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific™ 5-Bromo-7-chloro-1H-indazole, 96%
Thermo Scientific™ 5-Bromo-7-chloro-1H-indazole, 96%
CAS: 635712-44-6 Molecular Formula: C{7}H{4}BrClN{2} MDL Number: MFCD11044588