accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6358-09-4

IUPAC Name: 3-chloro-2-hydroxy-5-nitroanilinium chloride
Synonyms: 2-amino-6-chloro-4-nitrophenol
Molecular Weight (g/mol): 225.03
Molecular Formula: C6H6Cl2N2O3
InChi Key: HXKQPFKBBNRPEK-UHFFFAOYSA-N
SMILES: [Cl-].[NH3+]C1=CC(=CC(Cl)=C1O)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results