accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6358-64-1

IUPAC Name: 4-chloro-2,5-dimethoxyaniline
Synonyms: 4-chloro-2,5-dimethoxyaniline
Molecular Weight (g/mol): 187.62
Molecular Formula: C8H10ClNO2
InChi Key: YGUFQYGSBVXPMC-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=C(OC)C=C1N
Percent Purity 
  • (1)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)