accessibility menu, dialog, popup

UserName

Viewing profile as user:
SNH

CAS RN 636-04-4

IUPAC Name: N-phenylbenzenecarbothioamide
Synonyms: N-phenylbenzenecarbothioamide
Molecular Weight (g/mol): 213.3
Molecular Formula: C13H11NS
InChi Key: BOQKCADLPNLYCZ-UHFFFAOYSA-N
SMILES: S=C(NC1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results