accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 636-80-6

IUPAC Name: hydrogen pyridine-2-carboxylic acid chloride
Synonyms: hydrogen picolinic acid chloride
Molecular Weight (g/mol): 159.57
Molecular Formula: C6H6ClNO2
InChi Key: HFAFXVOPGDBAOK-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC(=O)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results