accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 636-82-8

IUPAC Name: cyclohex-1-ene-1-carboxylic acid
Synonyms: 1-cyclohexenecarboxylic acid
Molecular Weight (g/mol): 126.16
Molecular Formula: C7H10O2
InChi Key: NMEZJSDUZQOPFE-UHFFFAOYSA-N
SMILES: OC(=O)C1=CCCCC1
Boiling Point 
  • (1)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (2)
  • (4)
  • (3)
  • (1)
  • (2)
  • (2)