accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 636-95-3

IUPAC Name: hydrogen (3-nitrophenyl)hydrazine chloride
Synonyms: hydrogen (3-nitrophenyl)hydrazine chloride
Molecular Weight (g/mol): 189.6
Molecular Formula: C6H8ClN3O2
InChi Key: BKOYKMLGFFASBG-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=CC(=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)