accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH3CHNNOOOOCH3CH3CH3

CAS RN 63675-72-9

IUPAC Name: 3-methyl 5-(2-methylpropyl) 2,6-dimethyl-4-(2-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Synonyms: nisoldipine
Molecular Weight (g/mol): 388.42
Molecular Formula: C20H24N2O6
InChi Key: VKQFCGNPDRICFG-UHFFFAOYNA-N
SMILES: COC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC=C1[N+]([O-])=O)C(=O)OCC(C)C
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)