accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 637-39-8

IUPAC Name: hydrogen 2-[bis(2-hydroxyethyl)amino]ethan-1-ol chloride
Synonyms: tea-hydrochloride
Molecular Weight (g/mol): 185.65
Molecular Formula: C6H16ClNO3
InChi Key: HHLJUSLZGFYWKW-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OCCN(CCO)CCO
Molecular Weight (g/mol) 
  • (7)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
  • (5)
Physical Form 
  • (1)
  • (5)
  • (3)
Grade 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (5)
  • (1)
  • (3)
  • (7)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)
  • (1)
  • (5)
  • (3)
  • (3)
  • (5)