accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 637-53-6

CH3SNH

CAS RN 637-53-6

IUPAC Name: N-phenylethanethioamide
Synonyms: thioacetanilide
Molecular Weight (g/mol): 151.23
Molecular Formula: C8H9NS
InChi Key: MWCGLTCRJJFXKR-UHFFFAOYSA-N
SMILES: CC(=S)NC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results