accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClNO

CAS RN 637-61-6

IUPAC Name: 4-(chloroimino)cyclohexa-2,5-dien-1-one
Synonyms: quinone chloroimide
Molecular Weight (g/mol): 141.55
Molecular Formula: C6H4ClNO
InChi Key: ITUYMTWJWYTELW-UHFFFAOYSA-N
SMILES: ClN=C1C=CC(=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results