accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOO

CAS RN 637-64-9

IUPAC Name: (oxolan-2-yl)methyl acetate
Synonyms: tetrahydrofurfuryl acetate
Molecular Weight (g/mol): 144.17
Molecular Formula: C7H12O3
InChi Key: AAQDYYFAFXGBFZ-UHFFFAOYNA-N
SMILES: CC(=O)OCC1CCCO1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)