accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 637-65-0

H3COOO

CAS RN 637-65-0

IUPAC Name: (oxolan-2-yl)methyl propanoate
Synonyms: oxolan-2-ylmethyl propanoate
Molecular Weight (g/mol): 158.2
Molecular Formula: C8H14O3
InChi Key: FMKCDSXOYLTWBR-UHFFFAOYNA-N
SMILES: CCC(=O)OCC1CCCO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)