accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6372-40-3

IUPAC Name: diphenyl(propan-2-yl)phosphane
Synonyms: isopropyldiphenylphosphane
Molecular Weight (g/mol): 228.28
Molecular Formula: C15H17P
InChi Key: LLZAIAIZAVMQIG-UHFFFAOYSA-N
SMILES: CC(C)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)