CAS RN 6375-47-9
CAS RN 6375-47-9
IUPAC Name:
N-(3-amino-4-methoxyphenyl)acetamide
Synonyms:
N-(3-amino-4-methoxyphenyl)acetamide
Molecular Weight (g/mol):
180.21
Molecular Formula:
C9H12N2O2
InChi Key:
SJWQCBCAGCEWCV-UHFFFAOYSA-N
SMILES:
COC1=CC=C(NC(C)=O)C=C1N
1
–
1
of
1
results
1
3'-Amino-4'-methoxyacetanilide 98.0+%, TCI America™
CAS: 6375-47-9 Molecular Formula: C9H12N2O2 Molecular Weight (g/mol): 180.207 MDL Number: MFCD00008676 InChI Key: SJWQCBCAGCEWCV-UHFFFAOYSA-N Synonym: 5-Acetamido-o-anisidine, N1-Acetyl-4-methoxy-1,3-phenylenediamine PubChem CID: 80779 IUPAC Name: N-(3-amino-4-methoxyphenyl)acetamide SMILES: CC(=O)NC1=CC(=C(C=C1)OC)N