CAS RN 63758-12-3
IUPAC Name:
2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride
Synonyms:
2,3-dihydro-1,4-benzodioxine-6-sulfonyl chloride
Molecular Weight (g/mol):
234.65
Molecular Formula:
C8H7ClO4S
InChi Key:
JWHSRWUHRLYAPM-UHFFFAOYSA-N
SMILES:
ClS(=O)(=O)C1=CC2=C(OCCO2)C=C1
1
–
1
of
1
results
1
Sigma Aldrich 1,4-Benzodioxan-6-sulfonyl chloride
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.