accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 638-08-4

IUPAC Name: octadecanoyl octadecanoate
Synonyms: octadecanoic acid, anhydride
Molecular Weight (g/mol): 550.95
Molecular Formula: C36H70O3
InChi Key: WVJVHUWVQNLPCR-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)