accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOH3CH3C

CAS RN 638-10-8

IUPAC Name: ethyl 3-methylbut-2-enoate
Synonyms: ethyl 3-methylbut-2-enoate
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: UTXVCHVLDOLVPC-UHFFFAOYSA-N
SMILES: CCOC(=O)C=C(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results