CAS RN 638-21-1
IUPAC Name:
phenylphosphane
Synonyms:
phenylphosphine
Molecular Weight (g/mol):
110.1
Molecular Formula:
C6H7P
InChi Key:
RPGWZZNNEUHDAQ-UHFFFAOYSA-N
SMILES:
PC1=CC=CC=C1
1
–
2
of
2
results
2
Sigma Aldrich Pyrocatechol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.