accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNH2O

CAS RN 638-58-4

IUPAC Name: tetradecanamide
Synonyms: tetradecanamide
Molecular Weight (g/mol): 227.39
Molecular Formula: C14H29NO
InChi Key: QEALYLRSRQDCRA-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(N)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results