accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 638-65-3

H3CN

CAS RN 638-65-3

IUPAC Name: octadecanenitrile
Synonyms: octadecanenitrile
Molecular Weight (g/mol): 265.49
Molecular Formula: C18H35N
InChi Key: RHSBIGNQEIPSCT-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)