accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 638-79-9

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5-undecafluoro-5-iodopentane
Synonyms: pentane, undecafluoro-5-iodo-
Molecular Weight (g/mol): 395.94
Molecular Formula: C5F11I
InChi Key: KCEJJSGJNCSQFI-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results