accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OCaOSNOOOSNOO

CAS RN 6381-91-5

IUPAC Name: calcium bis(1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate) hydrate
Synonyms: calcium bis(1,3-dioxo-1λ⁶,2-benzothiazol-1-olate) hydrate
Molecular Weight (g/mol): 422.44
Molecular Formula: C14H10CaN2O7S2
InChi Key: XOCXGVIOTKFUSU-UHFFFAOYSA-L
SMILES: O.[Ca++].[O-]S1(=O)=NC(=O)C2=CC=CC=C12.[O-]S1(=O)=NC(=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)