CAS RN 6381-92-6
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline Powder/USP/EP/BP/JP), Fisher Chemical™
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Buffer Solution, pH 10.00 (Certified), Fisher Chemical™
Potassium Carbonate/Potassium Tetraborate/Potassium Hydroxide/Disodium EDTA Dihydrate
EDTA, Certified, 0.0500M ±0.0005M (0.1N), LabChem™
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Edetate Disodium, U.S.P., J.T. Baker™
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
EDTA Titrant, 0.0575 M, Ricca Chemical
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
EDTA Titrant, 0.100 M (M/10), Ricca Chemical
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
EDTA Titrant, 0.0500 M (M/20), Ricca Chemical
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
EDTA Titrant, 0.0100 M (M/100), Ricca Chemical
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic acid disodium salt dihydrate, 99+%
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic acid, (EDTA), 0.5M Solution, Molecular Biology Grade, Ultrapure
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.238 MDL Number: MFCD00150037 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate SMILES: C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, ACS reagent
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, disodium salt dihydrate, 99+%, for analysis
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic Acid, Disodium Salt, Standard Solution (0.1M/Certified), Fisher Chemical
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
Ethylenediaminetetraacetic Acid, Disodium Salt Dihydrate (Crystalline Powder/USP/FCC), Fisher Chemical™
CAS: 6381-92-6 Molecular Formula: C10H18N2Na2O10 Molecular Weight (g/mol): 372.24 MDL Number: MFCD00150037,MFCD00003541 InChI Key: OVBJJZOQPCKUOR-UHFFFAOYSA-L Synonym: edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs PubChem CID: 44120005 IUPAC Name: disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate SMILES: O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O