accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OKNH2(S)OOOHO

CAS RN 6382-01-0

IUPAC Name: potassium (4S)-4-amino-4-carboxybutanoate hydrate
Synonyms: potassium (4S)-4-amino-4-carboxybutanoate hydrate
Molecular Weight (g/mol): 203.24
Molecular Formula: C5H10KNO5
InChi Key: XIBUKSQTWSKJMQ-QTNFYWBSSA-M
SMILES: O.[K+].N[C@@H](CCC([O-])=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)