accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6382-93-0

IUPAC Name: (2R)-5-{[amino(iminiumyl)methyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoate
Synonyms: (2R)-5-{[amino(iminio)methyl]amino}-2-{[(benzyloxy)carbonyl]amino}pentanoate
Molecular Weight (g/mol): 308.34
Molecular Formula: C14H20N4O4
InChi Key: SJSSFUMSAFMFNM-LLVKDONJSA-N
SMILES: NC(=[NH2+])NCCC[C@@H](NC(=O)OCC1=CC=CC=C1)C([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results