CAS RN 63843-46-9
IUPAC Name:
4-(3-methylphenoxy)piperidine
Synonyms:
4-(3-methylphenoxy)piperidine
Molecular Weight (g/mol):
191.27
Molecular Formula:
C12H17NO
InChi Key:
RTSXWJCYCFVWBU-UHFFFAOYSA-N
SMILES:
CC1=CC(OC2CCNCC2)=CC=C1
1
–
1
of
1
results
1
Promotions Available
4-(3-Methylphenoxy)piperidine, 97%, Thermo Scientific Chemicals
4-(3-Methylphenoxy)piperidine, 97%, Thermo Scientific Chemicals
CAS: 63843-46-9 Molecular Formula: C12H17NO Molecular Weight (g/mol): 191.274 MDL Number: MFCD06248345 InChI Key: RTSXWJCYCFVWBU-UHFFFAOYSA-N Synonym: 4-3-methylphenoxy piperidine,4-m-tolyloxy piperidine,acmc-1b64o PubChem CID: 17196730 IUPAC Name: 4-(3-methylphenoxy)piperidine SMILES: CC1=CC(=CC=C1)OC2CCNCC2