accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6385-02-0

IUPAC Name: sodium 2-[(2,6-dichloro-3-methylphenyl)amino]benzoate
Synonyms: sodium 2-[(2,6-dichloro-3-methylphenyl)amino]benzoate
Molecular Weight (g/mol): 318.13
Molecular Formula: C14H10Cl2NNaO2
InChi Key: OGPIIGMUPMPMNT-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=C(Cl)C(NC2=CC=CC=C2C([O-])=O)=C1Cl
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (5)
Physical Form 
  • (1)
  • (5)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (5)
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)