accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6388-47-2

IUPAC Name: 2-amino-3-chlorobenzoic acid
Synonyms: 2-amino-3-chlorobenzoic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: LWUAMROXVQLJKA-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=CC=C1C(O)=O
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results