accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63884-78-6

H3CONH2

CAS RN 63884-78-6

IUPAC Name: 1-(4-aminophenyl)octan-1-one
Synonyms: 4-aminooctoylphenone
Molecular Weight (g/mol): 219.33
Molecular Formula: C14H21NO
InChi Key: IMQPGJKTCYTPEI-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)C1=CC=C(N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results