accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONHNCl

CAS RN 63910-43-0

IUPAC Name: 4-chloro-5-methoxy-2,3-dihydropyridazin-3-one
Synonyms: 4-chloro-5-methoxy-2H-pyridazin-3-one
Molecular Weight (g/mol): 160.56
Molecular Formula: C5H5ClN2O2
InChi Key: SUZFURZANOKZBL-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C(=O)NN=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results