accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63927-22-0

IUPAC Name: 8-bromoisoquinoline
Synonyms: 8-bromoisoquinoline
Molecular Weight (g/mol): 208.06
Molecular Formula: C9H6BrN
InChi Key: DPRIHFQFWWCIGY-UHFFFAOYSA-N
SMILES: BrC1=C2C=NC=CC2=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)