accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2CH3H2N

CAS RN 6393-01-7

IUPAC Name: 2,5-dimethylbenzene-1,4-diamine
Synonyms: 4-amino-2,5-dimethylaniline
Molecular Weight (g/mol): 136.2
Molecular Formula: C8H12N2
InChi Key: BWAPJIHJXDYDPW-UHFFFAOYSA-N
SMILES: CC1=CC(N)=C(C)C=C1N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results