accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63959-45-5

IUPAC Name: (3R,4S,5R)-4,5-dihydroxy-3-(phosphonooxy)cyclohex-1-ene-1-carboxylic acid
Synonyms: 3-phosphoshikimic acid
Molecular Weight (g/mol): 254.13
Molecular Formula: C7H11O8P
InChi Key: QYOJSKGCWNAKGW-PBXRRBTRSA-N
SMILES: O[C@@H]1CC(=C[C@@H](OP(O)(O)=O)[C@H]1O)C(O)=O
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results