accessibility menu, dialog, popup

UserName

Viewing profile as user:
IPI

CAS RN 6396-07-2

IUPAC Name: diiodotriphenyl-λ⁵-phosphane
Synonyms: diiodotriphenyl-λ⁵-phosphane
Molecular Weight (g/mol): 516.1
Molecular Formula: C18H15I2P
InChi Key: NSWZEXJQHIXCFL-UHFFFAOYSA-N
SMILES: IP(I)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results