accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NH2ONHOHNNH2NH2ONHOHNOHSOHOO

CAS RN 63981-35-1

IUPAC Name: bis(5,6-diamino-1,2,3,4-tetrahydropyrimidine-2,4-dione); sulfuric acid
Synonyms: bis(5,6-diaminouracil); sulfuric acid
Molecular Weight (g/mol): 382.31
Molecular Formula: C8H14N8O8S
InChi Key: OFZPGHXQOXIBIV-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=C(N)C(=O)NC(=O)N1.NC1=C(N)C(=O)NC(=O)N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)