accessibility menu, dialog, popup

UserName

Viewing profile as user:
PBrHPhPhPh

CAS RN 6399-81-1

IUPAC Name: triphenylphosphane hydrobromide
Synonyms: triphenylphosphine hydrobromide
Molecular Weight (g/mol): 343.2
Molecular Formula: C18H16BrP
InChi Key: CMSYDJVRTHCWFP-UHFFFAOYSA-N
SMILES: Br.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)