accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONHO

CAS RN 64-65-3

IUPAC Name: 4-ethyl-4-methylpiperidine-2,6-dione
Synonyms: 3-ethyl-3-methylglutarimide bemegride (inn) and its salts
Molecular Weight (g/mol): 155.2
Molecular Formula: C8H13NO2
InChi Key: ORRZGUBHBVWWOP-UHFFFAOYSA-N
SMILES: CCC1(C)CC(=O)NC(=O)C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results