accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64-69-7

IUPAC Name: 2-iodoacetic acid
Synonyms: iodoacetic acid
Molecular Weight (g/mol): 185.95
Molecular Formula: C2H3IO2
InChi Key: JDNTWHVOXJZDSN-UHFFFAOYSA-N
SMILES: OC(=O)CI
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)