accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 640-57-3

IUPAC Name: 1-(benzenesulfonyl)-4-methylbenzene
Synonyms: 1-(benzenesulfonyl)-4-methylbenzene
Molecular Weight (g/mol): 232.3
Molecular Formula: C13H12O2S
InChi Key: YBRXHRWLEFCFEG-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results