accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64013-70-3

IUPAC Name: 2-(aminomethyl)-6-[(4,6-diamino-3-{[4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2-hydroxycyclohexyl)oxy]oxane-3,4,5-triol; bis(sulfuric acid)
Synonyms: kanamycin; bis(sulfuric acid)
Molecular Weight (g/mol): 680.65
Molecular Formula: C18H40N4O19S2
InChi Key: OGTKIXVMLDAMNU-UHFFFAOYNA-N
SMILES: OS(O)(=O)=O.OS(O)(=O)=O.NCC1OC(OC2C(N)CC(N)C(OC3OC(CO)C(O)C(N)C3O)C2O)C(O)C(O)C1O
Color 
  • (1)
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results