accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 64019-57-4

IUPAC Name: tris[(1H-1,3-benzodiazol-2-yl)methyl]amine
Synonyms: tris(1H-1,3-benzodiazol-2-ylmethyl)amine
Molecular Weight (g/mol): 407.48
Molecular Formula: C24H21N7
InChi Key: YQIGEJHOYBUSLR-UHFFFAOYSA-N
SMILES: C(N(CC1=NC2=CC=CC=C2N1)CC1=NC2=CC=CC=C2N1)C1=NC2=CC=CC=C2N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results