accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6402-09-1

IUPAC Name: 3-methyl-1-(4-nitrophenyl)-4,5-dihydro-1H-pyrazol-5-one
Synonyms: 5-methyl-2-(4-nitrophenyl)-4H-pyrazol-3-one
Molecular Weight (g/mol): 219.2
Molecular Formula: C10H9N3O3
InChi Key: MYPAMGFTHVEING-UHFFFAOYSA-N
SMILES: CC1=NN(C(=O)C1)C1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results