accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHO(E)

CAS RN 6402-36-4

IUPAC Name: (2E)-dodec-2-enedioic acid
Synonyms: traumatic acid dodec-2-enedioic acid traumatate trans-traumatate dodec-2c-enedioic acid dodec-2t-enedioate dodec-2c-enedioate dodec-2t-enedioic acid
Molecular Weight (g/mol): 228.29
Molecular Formula: C12H20O4
InChi Key: MAZWDMBCPDUFDJ-VQHVLOKHSA-N
SMILES: OC(=O)CCCCCCCC\C=C\C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results