accessibility menu, dialog, popup

UserName

Viewing profile as user:
NHOHN

CAS RN 64030-43-9

IUPAC Name: N-phenylpyrrolidine-2-carboxamide
Synonyms: N-phenylpyrrolidine-2-carboxamide
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: NRXYBKZDRCXDOD-UHFFFAOYNA-N
SMILES: O=C(NC1=CC=CC=C1)C1CCCN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)