accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6404-26-8

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-6-acetamidohexanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]-6-acetamidohexanoic acid
Molecular Weight (g/mol): 288.34
Molecular Formula: C13H24N2O5
InChi Key: IOKOUUAPSRCSNT-UEQNJFAPNA-N
SMILES: CC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results