accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(S)HNOHOOOCH3CH3CH3

CAS RN 6404-28-0

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: (2S)-2-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 231.29
Molecular Formula: C11H21NO4
InChi Key: ZIOCIQJXEKFHJO-QMMMGPOBSA-N
SMILES: CCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results