CAS RN 64048-12-0
IUPAC Name:
4-[3,4,5-tris(pyridin-4-yl)thiophen-2-yl]pyridine
Synonyms:
4-[3,4,5-tris(pyridin-4-yl)thiophen-2-yl]pyridine
Molecular Weight (g/mol):
392.48
Molecular Formula:
C24H16N4S
InChi Key:
USWLOKMMUTWFMD-UHFFFAOYSA-N
SMILES:
S1C(=C(C(=C1C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1)C1=CC=NC=C1
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Hh/Gli Antagonist, GANT58
The Hh/Gli Antagonist, GANT58, also referenced under CAS 64048-12-0, controls the biological activity of Hh/Gli. This small molecule/inhibitor is primarily used for Cell Signaling applications.