accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClOHHN

CAS RN 64051-79-2

IUPAC Name: hydrogen piperidin-3-ol chloride
Synonyms: hydrogen piperidin-3-ol chloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: VLECDMDGMKPUSK-UHFFFAOYNA-N
SMILES: [H+].[Cl-].OC1CCCNC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results